Anti-DNAJC22 Rabbit Polyclonal Antibody

Supplier: Abgent
AP10871B
89378-002EA 619.21 USD
89378-002
Anti-DNAJC22 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAJC22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR