Anti-RNASET2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP10764A
89377-762EA 619.55 USD
89377-762
Anti-RNASET2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNASET2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR