Anti-MB21D1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP10510C
89377-210EA 619.42 USD
89377-210
Anti-MB21D1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MB21D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR