Anti-LIPI Rabbit Polyclonal Antibody

Supplier: Abgent
AP10266B
89376-660EA 589.74 USD
89376-660
Anti-LIPI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIPI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR