Anti-EPHB3 Mouse Monoclonal Antibody [clone: 50CT97.1.9]

Supplier: Abgent
AM7624A
89376-020EA 590.05 USD
89376-020
Anti-EPHB3 Mouse Monoclonal Antibody [clone: 50CT97.1.9]
Antibodies
Type: Primary
Antigen: EPHB3
Clonality: Monoclonal
Clone: 50CT97.1.9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR