Anti-EIF4EBP1 Mouse Monoclonal Antibody [clone: 319CT22.8.2]

Supplier: Abgent
AM1938A
89375-958EA 590.05 USD
89375-958
Anti-EIF4EBP1 Mouse Monoclonal Antibody [clone: 319CT22.8.2]
Antibodies
Type: Primary
Antigen: EIF4EBP1
Clonality: Monoclonal
Clone: 319CT22.8.2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR