Anti-GNPDA1 Mouse Monoclonal Antibody [clone: 237CT2.5.2]

Supplier: Abgent
AM1910B
89375-940EA 619.55 USD
89375-940
Anti-GNPDA1 Mouse Monoclonal Antibody [clone: 237CT2.5.2]
Antibodies
Type: Primary
Antigen: GNPDA1
Clonality: Monoclonal
Clone: 237CT2.5.2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human (Predicted), Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR