Anti-GBL Mouse Monoclonal Antibody [clone: 211CT7.1.1]

Supplier: Abgent
AM1904A
89375-922EA 619.55 USD
89375-922
Anti-GBL Mouse Monoclonal Antibody [clone: 211CT7.1.1]
Antibodies
Type: Primary
Antigen: GBL
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human (Predicted), Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR