Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 225CT3.1.3]

Supplier: Abgent
AM1898B
89375-906EA 619.55 USD
89375-906
Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 225CT3.1.3]
Antibodies
Type: Primary
Antigen: EIF4E2
Clonality: Monoclonal
Clone: 225CT3.1.3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR