Anti-DPF2 Mouse Monoclonal Antibody [clone: 226CT14.1.1.2]

Supplier: Abgent
AM1897A
89375-900EA 619.55 USD
89375-900
Anti-DPF2 Mouse Monoclonal Antibody [clone: 226CT14.1.1.2]
Antibodies
Type: Primary
Antigen: DPF2
Clonality: Monoclonal
Clone: 226CT14.1.1.2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR