Anti-TGFB2 Mouse Monoclonal Antibody [clone: 220ct16.4.3.1]

Supplier: Abgent
AM1893A
89375-884EA 590.05 USD
89375-884
Anti-TGFB2 Mouse Monoclonal Antibody [clone: 220ct16.4.3.1]
Antibodies
Type: Primary
Antigen: TGFB2
Clonality: Monoclonal
Clone: 220ct16.4.3.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR