Anti-ULK2 Mouse Monoclonal Antibody [clone: 218CT21.2.6]

Supplier: Abgent
AM1884A
89375-852EA 590.05 USD
89375-852
Anti-ULK2 Mouse Monoclonal Antibody [clone: 218CT21.2.6]
Antibodies
Type: Primary
Antigen: ULK2
Clonality: Monoclonal
Clone: 218CT21.2.6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR