Anti-URM1 Mouse Monoclonal Antibody [clone: 182CT27.2.2]

Supplier: Abgent
AM1883A
89375-850EA 619.55 USD
89375-850
Anti-URM1 Mouse Monoclonal Antibody [clone: 182CT27.2.2]
Antibodies
Type: Primary
Antigen: URM1
Clonality: Monoclonal
Clone: 182CT27.2.2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR