Anti-HSPA1A Mouse Monoclonal Antibody [clone: 201CT8.2.3]

Supplier: Abgent
AM1877A
89375-832EA 590.05 USD
89375-832
Anti-HSPA1A Mouse Monoclonal Antibody [clone: 201CT8.2.3]
Antibodies
Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 201CT8.2.3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR