Anti-UBTD1 Mouse Monoclonal Antibody [clone: 189CT14.5.4]

Supplier: Abgent
AM1873A
89375-818EA 590.05 USD
89375-818
Anti-UBTD1 Mouse Monoclonal Antibody [clone: 189CT14.5.4]
Antibodies
Type: Primary
Antigen: UBTD1
Clonality: Monoclonal
Clone: 189CT14.5.4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR