Anti-GFAP Mouse Monoclonal Antibody [clone: 183CT3.1.5]

Supplier: Abgent
AM1870A
89375-806EA 590.05 USD
89375-806
Anti-GFAP Mouse Monoclonal Antibody [clone: 183CT3.1.5]
Antibodies
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 183CT3.1.5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR