Anti-MAPK15 Mouse Monoclonal Antibody [clone: 168CT10.6.6]

Supplier: Abgent
AM1855A
89375-748EA 589.89 USD
89375-748
Anti-MAPK15 Mouse Monoclonal Antibody [clone: 168CT10.6.6]
Antibodies
Type: Primary
Antigen: MAPK15
Clonality: Monoclonal
Clone: 168CT10.6.6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR