Anti-MAP2 Mouse Monoclonal Antibody [clone: 159CT34.12.3.4]

Supplier: Abgent
AM1853B
89375-742EA 590.05 USD
89375-742
Anti-MAP2 Mouse Monoclonal Antibody [clone: 159CT34.12.3.4]
Antibodies
Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: 159CT34.12.3.4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR