Anti-EIF4E Mouse Monoclonal Antibody [clone: 163CT48.1.9]

Supplier: Abgent
AM1852A
89375-736EA 615.89 USD
89375-736
Anti-EIF4E Mouse Monoclonal Antibody [clone: 163CT48.1.9]
Antibodies
Type: Primary
Antigen: EIF4E
Clonality: Monoclonal
Clone: 163CT48.1.9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR