Anti-AR Mouse Monoclonal Antibody [clone: 130CT43.8.5]

Supplier: Abgent
AM1824A
89375-640EA 619.55 USD
89375-640
Anti-AR Mouse Monoclonal Antibody [clone: 130CT43.8.5]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 130CT43.8.5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR