LabHand Holder, Diversified Woodcrafts
Supplier: VWR International
89375-414
89375-414EA 367.06 USD
89375-414
LabHand Holder, Diversified Woodcrafts
Computer and Equipment Stands and Mounts
Designed to provide a safe, convenient surface for laptops, tablets or books in a laboratory setting.
  • Mounted on a solid plastic rod
  • Protects laptops and books from spills
  • Increases available workspace
The LabHand is mounted on a solid plastic rod which fits securely into standard rod sockets. The platform is made of silver colored steel to create a surface that will withstand the typical chemicals found in laboratories. The LabHand holds laptops and books off the work surface protecting them from spills and increasing the amount of workspace available. 12" square with 5¹/₂" rod.