VWR® Six-Student Single Faced Workstation with Science Center
Supplier: VWR International
89374-996
89374-996EA 26783.02 USD
89374-996
VWR® Six-Student Single Faced Workstation with Science Center
Furniture Tables and Workbenches
Six student single faced workstation that provides a fully equipped science center.
  • 1" solid epoxy resin surface
  • Constructed of solid oak and oak hardwood veneers
  • Includes six book compartments in the aprons
  • The cabinets have locking cupboard
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
The single faced units contain a solid epoxy resin sink along with cold water faucets for every two students. Every unit contains enough gas cocks for each student along with GFI AC duplex electrical receptacles, rod sockets and book compartments. All units contain locks for the drawers and doors. Choose from models with a full cupboard only or a half cupboard and dovetailed drawers. Can be ordered without sink, fixtures and rod sockets. They are finished with a chemical resistant, earth-friendly UV finish. Add 66" in length for each additional two students. Your choice of 1" epoxy resin or ¾" phenolic resin tops.
Sink dimension: 12W×8L×6"D
Certifications: SEFA compliant and MAS certified green.