ZetexPlus Heat Resistant Gloves with Double Palm
Supplier: Newtex Industries
2100017
89370-992PR 223.03 USD
89370-992
ZetexPlus Heat Resistant Gloves with Double Palm
Gloves
Protection from temperatures up to 1093 °C (2000 °F).
Common applications: Foundries and casting operations, welding and hot works operations, glass and ceramic plants production, power generation, petrochemical and refining, laboratory and testing facilities.
Woven from 100% premium ZetexPlus texturized fiberglass and treated with a proprietary vermiculite coating. The double palm allows for a longer product life and improved heat resistance. Double Stitched and reinforced. Premium wool liner. Available in 35.6 or 58.4 cm (14 or 23") lengths. 35.6 cm (14") cuff protects wrists. 58.4 cm (23") glove features a gauntlet style cuff. One size fits all. Made in the USA.
Certifications: EN 388, EN 407.