Hydrion® Quat Test Papers, Micro Essential Laboratory
Supplier: Micro Essential
QC-1001
89370-914CS 97.42 USD
89370-914
Hydrion® Quat Test Papers, Micro Essential Laboratory
Test Paper Indicator Paper
A quick, reliable, easy-to-use, way to test quaternary ammonium based sanitizers in food service establishments, as required by federal, state, and local food safety codes.
Delivery information: Kit consists of a 15' x 5/32" roll of premium test paper in a handy Hydrion dispenser with a matching color chart.