SHOWA 772 Nitrile, Chemical Resistant, 26" Glove, Showa
Supplier: Showa
772M-08 772L-09 772XL-10
89370-885CS 2967.66 USD
89370-885 89370-887 89370-889
SHOWA 772 Nitrile, Chemical Resistant, 26" Glove, Showa
Gloves
Nitrile chemical resistant gloves provides protection to the shoulder against a wide range of chemicals.
  • Highly flexible nitrile coating
  • Wide range of chemical protection
  • Cotton interlocked knit liner
The 26" length provides maximum protection up to the wearer's shoulder. Nitrile coating provides protection from snags, punctures, abrasions, and cuts.
Certifications: ANSI Abrasion Level 3.