Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Epredia™ Histoplast Plus
Supplier: Epredia
8331
89370-812CS
175.63
USD
89370-812
Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Epredia™ Histoplast Plus
Paraffin wax blended with synthetic polymers
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle–free sections
- Also available prepackaged with Epredia™ Excelsior™ AS and Revos™ tissue processor waste wax containers for ease of use
- Histoplast PE is good for general infiltration and embedding procedures (melting point 56°C)
- Histoplast IM utilizes DMSO to help infiltrate larger tissues samples (melting point 56°C)
- Histoplast LP has a lower melting point 50-54°C for small and friable tissue samples
CAS Number:
8002-74-2 |