Flask Clamp, Humboldt
Supplier: Humboldt Manufacturing
H-23442
89370-276EA 74.54 USD
89370-276
Flask Clamp, Humboldt
Clamps General Purpose Clamps
Flask clamps provide safe handling of flasks and stoppers sizes 4 to 8
Jaws are "V"-shaped and open up to a 48 mm diameter. Made of stainless steel with a riveted joint.