Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis), VWR® ACS, Reagent Grade for histology

Supplier: VWR International
Danger

Synonyms: Dimethylbenzene

ES34411VWR
89370-088CS 544.33 USD
89370-088
Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis), VWR® ACS, Reagent Grade for histology
Xylene (mixture of isomers)

VWR®, part of Avantor has industry leading Anatomical Pathology reagents available your lab.


  • ISO 13485 Certified
  • F-style gallons for easier storage and fewer spills
  • Transparency and availability to help you with reliability


Certifications: ISO 13485 Certified

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chemical Assay (GC), (Xylene Isomers & Ethyl Benzene) 98.5% min.
Paraxylene ( p-xylene) Reported per lot
Metaxylene (m-xylene) Reported per lot
Orthoxylene (o-xylene) Reported per lot
Ethyl Benzene 25% max.
Boiling Range 137-143 °C
Color (APHA) 10 max.
Appearance Clear
Residue After Evaporation 10 ppm max.
Substances Darkened by H₂SO₄ To Pass
Sulphur Compounds 1 ppm max.
Water 0.05% max.
Specific Gravity @ 15.56°C 0.865 - 0.875
Benzene Content 0.01% max.
Toluene Content 0.5% max.
Total C9 Aromatics Reported per lot
Non-Aromatics 0.30% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR