Alcohol 100%, VWR®, Reagent Grade for histology

Supplier: VWR International
Danger

Synonyms: Reagent alcohol

ES34116VWR
89370-084CS 111.82 USD
89370-084
Alcohol 100%, VWR®, Reagent Grade for histology
Alcohol

Reagent Grade Alcohol, 100%


VWR Histology Reagents are manufactured from high quality raw materials in an ISO 13485 facility, ensuringoptimal performance in your histology lab.

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Components
E/E 89.5-91.5%
Methanol 4.0-5.0%
Isopropanol 4.5-5.5%
Characteristics
Methanol and Ethyl Alcohol (SDA 3A 200 Proof) (v/v) 94.0 – 96.0%
Isopropyl Alcohol (2-Propanol) (v/v) 4.0 – 6.0%
Water max 0.2%
Residue After Evaporation max 10 ppm
Appearance Clear
Specific Gravity 0.7902-0.7912 @ 20° C
Color (Pt-Co) 10 max
Odor Pass
Titrable Acid 0.0003meq/g
Titrable Base 0.0002meq/g
Flourescent Background Pass
Identification Pass
Substances Reducing KMnO4 Pass
Solubility in Water Pass
Refractive Index @ 25°C 1.3580-1.3610

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR