Alcohol 95%, VWR®, Reagent Grade for histology

Supplier: VWR International
Danger

89370-082
89370-082CS 103.07 USD
89370-082
Alcohol 95%, VWR®, Reagent Grade for histology
Alcohol

Reagent Grade Alcohol, 95%


VWR Histology Reagents are manufactured from high quality raw materials in an ISO 13485 facility, ensuringoptimal performance in your histology lab.

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Components
Ethanol – 190 Proof 89.5-91.5%
Methanol 4.0-5.0%
Isopropanol 4.5-5.5%
Characteristics
Methanol and Ethyl Alcohol (SDA 3A 190 proof) (v/v) 94.0-96.0%
Isopropyl Alcohol (2-Propanol) (v/v) 4.0-6.0%
Specific Gravity @20°C 0.810-0.812
Apparent Proof @20°C Lot Analysis
Color (Pt-Co) 10 max.
Acidity To Pass
Odor Characteristic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR