Alcohol 80%, VWR®, Reagent Grade for histology

Supplier: VWR International
Danger

89370-080
89370-080CS 102.19 USD
89370-080
Alcohol 80%, VWR®, Reagent Grade for histology
Alcohol

Reagent Grade Alcohol, 80%


VWR Histology Reagents are manufactured from high quality raw materials in an ISO 13485 facility, ensuringoptimal performance in your histology lab.

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Components
Ethanol 72.4%
Methanol 3.6%
Isopropanol 4%
Water 20.0%
Characteristics
Specific gravity @20°C 0.855-0.865
Appearance Clear liquid, free from sediment
Color (Pt-Co) 10 max.
Acidity (wt/wt as acetic acid) 0.0025%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR