Alcohol 70%, VWR®, Reagent Grade for histology

Supplier: VWR International
Danger

Synonyms: Reagent alcohol

ES34110VWR
89370-078CS 110.82 USD
89370-078
Alcohol 70%, VWR®, Reagent Grade for histology
Alcohol

VWR®, part of Avantor has industry leading Anatomical Pathology reagents available your lab.


  • ISO 13485 Certified
  • F-style gallons for easier storage and fewer spills
  • Transparency and availability to help you with reliability


Certifications: ISO 13485 Certified

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Components
E/E 63.3%
Methanol 3.2%
Isopropanol 3.5%
Water 30.0%
Characteristics
Specific gravity @20°C 0.880-0.890
Appearance Clear liquid, free from sediment
Color (Pt-Co) 10 max.
Acidity (wt/wt as acetic acid) 0.0025%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR