VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
Supplier: VWR International
90013-45 VC6404
89369-158EA 630.11 USD
89369-158 10754-722 10754-842
VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
Hygrometers Digital Thermo-Hygrometers
Traceable® Memory-Card Humidity/Temperature/Dew Point meter is a stand-alone unit with data logging capabilities.
  • Memory feature stores minimum/maximum readings
  • Visual and audio alarm features
  • Bright LED display
  • Unique 4-line display
It records readings from once a minute up to once every 720 minutes on a removable standard SD memory card
Minimum/maximum feature displays highest and lowest readings for any time period. Alarm feature sounds and flashes a bright red LED when temperature, humidity and/or dew points rise above or fall below user-set trip points. Alarm sets in 0.1 increments. Unique 4-line display simultaneously shows ambient temperature, humidity, dew point and time of day.
Memory card reader (supplied) plugs into any USB computer port for an instant report. Text report includes date/time of day and readings of sensors and can be sent to any spreadsheet or database. Unit monitors cabinets, incubators, clean rooms, store rooms, labs, hoods, and critical areas. Unit captures 5.9 million readings with the supplied 256 mb removable SD card.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with probe, cable, 256 mb SD memory card, USB memory card reader, magnetic mounting strips, Velcro®, wall mount, bench stand, batteries, adaptor, Traceable® certificate.