VWR® Traceable® Platinum Ultra-Accurate Digital Reference Thermometer
Supplier: VWR International
89369-138
89369-138EA 775.17 USD
89369-138
VWR® Traceable® Platinum Ultra-Accurate Digital Reference Thermometer
Thermometers Digital Thermometers
High-precision unit is designed for years of reliable service, even in the most severe environments.
  • Highly precise
  • Membrane keys are splash-proof and easy to operate
  • Instant response probe
  • Hold button freezes display to retain data
  • Real-time readings
Platinum sensor insures linear response and high accuracy across the entire temperature range. Temperature range for this thermometer is -200.000° to 500.000°C (-328.000° to 932.000°F). Resolution is 0.001° or 0.0001°. Thermometer is ideal for pharmaceuticals, foods, petroleum products, soils, ovens and electronics manufacturing. Membrane keys are splash-proof and easy to operate. Decimal-placement key changes display to show one, two, three or four significant numbers after the decimal. Internal data logger automatically captures minimum and maximum readings every hour for the last 24 hours. Instant response probe updates readings 3 times per second for liquid, air/gas, semi-solids. Hold button allows user to freeze display for reading later. Computer output allows readings to be captured in real time.
Certifications: To ensure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibrated laboratory accredited by A2LA. It indicates traceability to NIST.
Ordering information: Supplied with a platinum probe/cable, plastic carrying case, battery and Traceable® Certificate.