Anti-ACT1 Mouse Monoclonal Antibody [clone: 3F1]

Supplier: Genetex
GTX18146
89367-494EA 758.4 USD
89367-494
Anti-ACT1 Mouse Monoclonal Antibody [clone: 3F1]
Antibodies
The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are constituents of many cell types and are involved in a myriad of cellular processes including locomotion, secretion, cytoplasmic streaming, phagocytosis and cytokinesis. Although actin is one of the most conserved eukaryotic proteins, it is expressed in mammals and birds as at least six isoforms characterized by electrophoresis and amino acid sequence analysis. Four of them represent the differentiation markers of muscle tissues and two are found in practically all cells.

IF: Use at an assay dependent dilution. IHC-P: Use at a concentration of 1 - 2 ?g/ml for 30 min at RT. No special pretreatment is required for immunohistochemical staining of formalin-fixed tissues. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR