Anti-CRF Chicken Polyclonal Antibody

Supplier: Genetex
GTX17475
89367-486EA 706.3 USD
89367-486
Anti-CRF Chicken Polyclonal Antibody
Antibodies
GTX17475 recognises Corticotropin Releasing Factor(CRF)precursor and CRF peptide.

For WB: Use at an assay dependent dilution. Detects a band of approximately 70 kDa due to conjugation of recombinant CRF antigen to fusion protein. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR