Anti-HIST1H1B Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87506
89366-926EA 629.01 USD
89366-926
Anti-HIST1H1B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Histone H1

We recommend the following starting dilutions: For WB: Use at a dilution of 1:500-1:1000. For IHC-P: Use at a dilution of 1:50-1:100. For ICC/IF: Use at a dilution of 1:100-1:500. For ELISA: Use at a dilution of 1:1000. Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HIST1H1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR