Anti-OX1R Rabbit Polyclonal Antibody

Supplier: Genetex
GTX78117
89366-508EA 539.28 USD
89366-508
Anti-OX1R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Orexin Receptor 1 (OX1R)

For paraffin sections, the antibody was used at 3.8 ug/ml.

Type: Primary
Antigen: OX1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR