Anti-S1PR3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77925
89366-424EA 629.21 USD
89366-424
Anti-S1PR3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Sphingolipid Receptor Edg3/S1P3

Type: Primary
Antigen: S1PR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR