Anti-S1PR1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77919
89366-420EA 629.21 USD
89366-420
Anti-S1PR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Sphingolipid Receptor Edg1/S1P1

For IHC-P: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: S1PR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR