Anti-CCRN4L Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77906
89366-406EA 632.37 USD
89366-406
Anti-CCRN4L Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to CCRN4L

Type: Primary
Antigen: CCRN4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR