Anti-ASGR2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77833
89366-336EA 648.17 USD
89366-336
Anti-ASGR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to ASGR2

For WB: Suggested dilution at 4.0 ug/ml in 5% skim milk/PBS buffer. For IHC-P: Use at 4-8?g/ml. Not yet tested in other applications. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: ASGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR