Anti-CLDN8 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77832
89366-334EA 632.37 USD
89366-334
Anti-CLDN8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Claudin-8 (CLDN8)

For IHC-P: Use at a concentration of 4-8 µg/ml. For WB: Use at a concentration of 0.5-1 ug/ml in 5% skim milk/PBS buffer. Not tested in other applications. Optimal dilutions should be determined by researcher.

Type: Primary
Antigen: CLDN8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR