Anti-APMAP Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77828
89366-326EA 632.37 USD
89366-326
Anti-APMAP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Adiponectin (ACDC)

For WB: Use at a concentration of 5 ?g/ml in 5% skim milk/PBS buffer. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: APMAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR