Anti-IRF9 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX77807
89366-310EA 629.94 USD
89366-310
Anti-IRF9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to ISGF3G

WB: Suggested dilution at 0.005-0.05 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: IRF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR