Anti-LIG1 Mouse Polyclonal Antibody

Supplier: Genetex
GTX70139
89364-706EA 595.14 USD
89364-706
Anti-LIG1 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal to DNA ligase I

Not tested in other applications. Optima dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: LIG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR