Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: B023]

Supplier: Genetex
GTX41184
89363-444EA 672.53 USD
89363-444
Anti-Respiratory Syncytial Virus Mouse Monoclonal Antibody [clone: B023]
Antibodies
Respiratory syncytial virus (RSV) is a major cause of respiratory illness in young children. RSV infection produces a variety of signs and symptoms involving different areas of the respiratory tract, from the nose to the lungs. RSV is a negative sense, enveloped RNA virus. The virion is variable in shape and size with average diameter of between 120 and 300 nm. The 63 kD RSV fusion protein of the RSS 2 strain (subtype A) directs fusion of viral and cellular membranes, results in viral penetration, and can direct fusion of infected cells with adjoining cells, resulting in the formation of syncytia or multi nucleated giant cells.

Type: Primary
Antigen: Respiratory Syncytial Virus
Clonality: Monoclonal
Clone: [B023]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR