Anti-APOA1 Mouse Monoclonal Antibody [clone: 1402]

Supplier: Genetex
GTX40411
89363-176EA 672.31 USD
89363-176
Anti-APOA1 Mouse Monoclonal Antibody [clone: 1402]
Antibodies
Mouse monoclonal antibody [1402] to Apolipoprotein A1

EIA: Use at an assay dependent dilution. Competitive RIA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR