Anti-Hepatitis C Core NS3 + NS4 Goat Polyclonal Antibody

Supplier: Genetex
GTX40324
89363-152EA 672.53 USD
89363-152
Anti-Hepatitis C Core NS3 + NS4 Goat Polyclonal Antibody
Antibodies
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.

Type: Primary
Antigen: Hepatitis C Core NS3 + NS4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR