Anti-NAK ATPASE Mouse Monoclonal Antibody

Supplier: Genetex
GTX30203
89362-890EA 609.87 USD
89362-890
Anti-NAK ATPASE Mouse Monoclonal Antibody
Antibodies
Na,K-ATPase is an integral membrane protein complex that hydrolyzes ATP to maintain the transmembrane gradients of Na+ and K+ found in most mammalian cells. The enzyme is comprised of an alpha and Beta subunit. The a-polypeptide has been shown to be the catalytically active subunit, whereas the Beta-polypeptide is glycosylated and appears to be necessary for the assembly and transport of the sodium pump to the plasma membrane.

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR